3-(Trifluoromethylthio)phenol 95%

Synonyms: 3-[(Trifluoromethyl)thio]phenol

BD305792-1G BD305792-5G BD305792-250MG
BLDPBD305792-1GEA 80.9 EUR
BLDPBD305792-1G BLDPBD305792-5G BLDPBD305792-250MG
3-(Trifluoromethylthio)phenol 95%
3-(Trifluoromethylthio)phenol

Storage condition: Sealed in dry, 2-8°C

Formula: C₇H₅F₃OS
MW: 194,1700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06248888
CAS Number: 3823-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR