5-Nitrothiophene-2-carbonyl chloride 95%

Synonyms: 5-Nitrothiophene-2-carbonyl Chloride

BD305796-250MG BD305796-100MG
BLDPBD305796-250MGEA 24.2 EUR
BLDPBD305796-250MG BLDPBD305796-100MG
5-Nitrothiophene-2-carbonyl chloride 95%
5-Nitrothiophene-2-carbonyl chloride

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₅H₂ClNO₃S
MW: 191,59 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09260988
CAS Number: 39978-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR