1-[4-(Diphenylmethyl)-1-piperazinyl]-3,3-diphenyl-1-propanone 98%

BD305805-250MG BD305805-100MG
BLDPBD305805-250MGEA 111 EUR
BLDPBD305805-250MG BLDPBD305805-100MG
1-[4-(Diphenylmethyl)-1-piperazinyl]-3,3-diphenyl-1-propanone 98%
1-[4-(Diphenylmethyl)-1-piperazinyl]-3,3-diphenyl-1-propanone

Storage condition: Sealed in dry, Room Temperature

Formula: C₃₂H₃₂N₂O
MW: 460,60 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03387795
CAS Number: 41332-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR