4-Chloromethylbenzophenone 95%

BD305814-10G BD305814-25G BD305814-1G BD305814-5G
BLDPBD305814-10GEA 93.5 EUR
BLDPBD305814-10G BLDPBD305814-25G BLDPBD305814-1G BLDPBD305814-5G
4-Chloromethylbenzophenone 95%
4-Chloromethylbenzophenone

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₄H₁₁ClO
MW: 230,69 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04039476
CAS Number: 42728-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR