[3,3'-Biindolinylidene]-2,2'-dione 98+%

BD305827-50MG BD305827-100MG BD305827-250MG
BLDPBD305827-50MGEA 53.6 EUR
BLDPBD305827-50MG BLDPBD305827-100MG BLDPBD305827-250MG
[3,3'-Biindolinylidene]-2,2'-dione 98+%
[3,3'-Biindolinylidene]-2,2'-dione

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₁₆H₁₀N₂O₂
MW: 262,26 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00423577
CAS Number: 476-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR