Purmorphamine 98%

Synonyms: 2-(1-Naphthoxy)-6-(4-morpholinoanilino)-9-cyclohexylpurine,9-Cyclohexyl-N-(4-morpholinophenyl)-2-(naphthalen-1-yloxy)-9H-purin-6-amine

BD305828-100MG
BLDPBD305828-100MGEA 234 EUR
BLDPBD305828-100MG
Purmorphamine 98%
Purmorphamine

Storage condition: Keep in dark place, Sealed in dry, Store in freezer, under -20°C

Formula: C₃₁H₃₂N₆O₂
MW: 520,637 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09037557
CAS Number: 483367-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR