(2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanoic acid 50% in water

Synonyms: Gluconic Acid (contains Gluconolactone)

BLDPBD305849-1000GEA 34.7 EUR
BLDPBD305849-1000G BLDPBD305849-100G BLDPBD305849-500G BLDPBD305849-25G
(2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanoic acid 50% in water
(2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanoic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₆H₁₂O₇
MW: 196,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00066366
CAS Number: 526-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR