Ethyl 4-formamidobenzoate 95%

BD305854-5G BD305854-10G BD305854-1G BD305854-25G
BLDPBD305854-5GEA 34.7 EUR
BLDPBD305854-5G BLDPBD305854-10G BLDPBD305854-1G BLDPBD305854-25G
Ethyl 4-formamidobenzoate 95%
Ethyl 4-formamidobenzoate

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₀H₁₁NO₃
MW: 193,199 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00021025
CAS Number: 5422-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR