(Ac-Cys-OH)₂ 97%

Synonyms: N,N'-Diacetyl-L-Cystine

BD305863-5G BD305863-250MG BD305863-1G BD305863-100MG
BLDPBD305863-5GEA 410 EUR
BLDPBD305863-5G BLDPBD305863-250MG BLDPBD305863-1G BLDPBD305863-100MG
(Ac-Cys-OH)₂ 97%
(Ac-Cys-OH)₂

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₀H₁₆N₂O₆S₂
MW: 324,38 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00152053
CAS Number: 5545-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR