Fexaramine 98%

Synonyms: Methyl (E)-3-(3-(N-((4'-(dimethylamino)-[1,1'-biphenyl]-4-yl)methyl)cyclohexanecarboxamido)phenyl)acrylate

BD305872-50MG BD305872-100MG BD305872-25MG
BLDPBD305872-50MGEA 250 EUR
BLDPBD305872-50MG BLDPBD305872-100MG BLDPBD305872-25MG
Fexaramine 98%
Fexaramine

Storage condition: Sealed in dry, 2-8°C

Formula: C₃₂H₃₆N₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD09971007
CAS Number: 574013-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR