N-(3-Chlorophenyl)picolinamide 99+%

BD305890-1G BD305890-50MG BD305890-100MG BD305890-250MG
BLDPBD305890-1GEA 881 EUR
BLDPBD305890-1G BLDPBD305890-50MG BLDPBD305890-100MG BLDPBD305890-250MG
N-(3-Chlorophenyl)picolinamide 99+%
N-(3-Chlorophenyl)picolinamide

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₁₂H₉ClN₂O
MW: 232,66 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00548412
CAS Number: 61350-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR