β-Methylstyrene (cis- and trans- mixture) 95% stabilised

Synonyms: 1-Propenylbenzene,β-Methylstyrene,1-Phenyl-1-propene

Stabilisation: Stabilised with 4-tert-butylpyrocatechol (TBC)

BD305905-5G BD305905-25G BD305905-1G
BLDPBD305905-5GEA 107 EUR
BLDPBD305905-5G BLDPBD305905-25G BLDPBD305905-1G
β-Methylstyrene (cis- and trans- mixture) 95% stabilised
beta-Methylstyrene (cis- and trans- mixture)

Storage condition: Sealed in dry, 2-8°C

Formula: C₉H₁₀
MW: 118,18 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00009280
CAS Number: 637-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR