3-Ethylbenzophenone 95%

BD305918-5G BD305918-1G BD305918-250MG
BLDPBD305918-5GEA 276 EUR
BLDPBD305918-5G BLDPBD305918-1G BLDPBD305918-250MG
3-Ethylbenzophenone 95%
3-Ethylbenzophenone

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₅H₁₄O
MW: 210,2711 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01662328
CAS Number: 66067-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR