Ethyl 2-bromo-2-ethylbutanoate 95%

BLDPBD305935-10GEA 119 EUR
BLDPBD305935-10G BLDPBD305935-250MG BLDPBD305935-25G BLDPBD305935-5G BLDPBD305935-1G
Ethyl 2-bromo-2-ethylbutanoate 95%
Ethyl 2-bromo-2-ethylbutanoate

Storage condition: Inert atmosphere, 2-8°C

Formula: C₈H₁₅BrO₂
MW: 223,10 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00017856
CAS Number: 6937-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR