2-Chloro-9-(tetrahydropyran-2-yl)adenine 95%

BD305973-250MG BD305973-100MG BD305973-1G
BLDPBD305973-250MGEA 257 EUR
BLDPBD305973-250MG BLDPBD305973-100MG BLDPBD305973-1G
2-Chloro-9-(tetrahydropyran-2-yl)adenine 95%
2-Chloro-9-(tetrahydropyran-2-yl)adenine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₁₀H₁₂ClN₅O
MW: 253,68 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12964278
CAS Number: 77111-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR