1-Chloro-2,4-dibromo-6-fluorobenzene 98%

Synonyms: 2-Chloro-3,5-dibromo-1-fluorobenzene,2-Chloro-1,5-dibromo-3-fluorobenzene,1,5-Dibromo-2-chloro-3-fluorobenzene

BD3060-5G BD3060-25G BD3060-100G
BLDPBD3060-5GEA 26.3 EUR
BLDPBD3060-5G BLDPBD3060-25G BLDPBD3060-100G
1-Chloro-2,4-dibromo-6-fluorobenzene 98%
1-Chloro-2,4-dibromo-6-fluorobenzene

Storage condition: Sealed in dry, Room Temperature

Formula: C₆H₂Br₂ClF
MW: 288,34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00142891
CAS Number: 202925-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR