Ethyl-4-amino-3-hydroxybenzoate 95%

Synonyms: Ethyl 4-Amino-3-hydroxybenzoate

BD306017-1G BD306017-5G BD306017-250MG
BLDPBD306017-1GEA 60.9 EUR
BLDPBD306017-1G BLDPBD306017-5G BLDPBD306017-250MG
Ethyl-4-amino-3-hydroxybenzoate 95%
Ethyl-4-amino-3-hydroxybenzoate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₉H₁₁NO₃
MW: 181,19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01320538
CAS Number: 87081-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR