N-Methyl Omeprazole 95%

BD306029-50MG BD306029-100MG
BLDPBD306029-50MGEA 160 EUR
BLDPBD306029-50MG BLDPBD306029-100MG
N-Methyl Omeprazole 95%
N-Methyl Omeprazole

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₈H₂₁N₃O₃S
MW: 359,44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19441777
CAS Number: 89352-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR