(2-Iodo-5-methoxyphenyl)boronic acid 95%

BD306032-1G BD306032-250MG
BLDPBD306032-1GEA 84 EUR
BLDPBD306032-1G BLDPBD306032-250MG
(2-Iodo-5-methoxyphenyl)boronic acid 95%
(2-Iodo-5-methoxyphenyl)boronic acid

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₇H₈BIO₃
MW: 277,852 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26793796
CAS Number: 89694-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR