CGK733 98%

Synonyms: 2,2-diphenyl-N-[2,2,2-trichloro-1-[(4-fluoro-3-nitrophenyl)carbamothioylamino]ethyl]acetamide,2,2-Diphenyl-N-(2,2,2-trichloro-1-(3-(4-fluoro-3-nitrophenyl)thioureido)ethyl)acetamide,CGK 733,CGK733

BD306043-50MG
BLDPBD306043-50MGEA 319 EUR
BLDPBD306043-50MG
CGK733 98%
CGK733

Storage condition: Sealed in dry, 2-8°C

Formula: C₂₃H₁₈Cl₃FN₄O₃S
MW: 555,84 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09038682
CAS Number: 905973-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR