PluriSIn 1 98%

Synonyms: 4-Pyridinecarboxylic acid 2-Phenylhydrazide,N'-Phenylisonicotinohydrazide,NSC 14613,Isonicotinic acid 2-Phenylhydrazide

BD306047-100MG
BLDPBD306047-100MGEA 161 EUR
BLDPBD306047-100MG
PluriSIn 1 98%
PluriSIn 1

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formula: C₁₂H₁₁N₃O
MW: 213,24 g/mol
Storage Temperature: Freezer
MDL Number: MFCD02344904
CAS Number: 91396-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR