1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine 98%

Synonyms: 1,2-Dihexadecanoyl-sn-glycero-3-phosphoethanolamine,Dipalmitoylphosphatidylethanolamine,2-Aminoethyl (R)-2,3-Bis(hexadecanoyloxy)propyl Hydrogen Phosphate,(2R)-3-(((2-Aminoethoxy)(hydroxy)phosphoryl)oxy)propane-1,2-diyl dipalmitate,L-DPPE

BD306054-250MG BD306054-1G BD306054-100MG
BLDPBD306054-250MGEA 60.9 EUR
BLDPBD306054-250MG BLDPBD306054-1G BLDPBD306054-100MG
1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine 98%
1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formula: C₃₇H₇₄NO₈P
MW: 691,97 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00036776
CAS Number: 923-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR