Pyrrole-2,3,5-tricarboxylic Acid 97%

BD306071-100MG BD306071-50MG
BLDPBD306071-100MGEA 231 EUR
BLDPBD306071-100MG BLDPBD306071-50MG
Pyrrole-2,3,5-tricarboxylic Acid 97%
Pyrrole-2,3,5-tricarboxylic Acid

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₇H₅NO₆
MW: 199,11 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09991933
CAS Number: 945-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR