4'-(Methylamino)-3'-nitroacetophenone 95+%

Synonyms: 1-(4-(Methylamino)-3-nitrophenyl)ethanone

BD307534-100MG BD307534-250MG BD307534-1G
BLDPBD307534-100MGEA 132 EUR
BLDPBD307534-100MG BLDPBD307534-250MG BLDPBD307534-1G
4'-(Methylamino)-3'-nitroacetophenone 95+%
4'-(Methylamino)-3'-nitroacetophenone

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₉H₁₀N₂O₃
MW: 194,1900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14584991
CAS Number: 18076-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR