4-Butylthiobenzamide 95%

BD314372-250MG BD314372-5G BD314372-100MG BD314372-1G
BLDPBD314372-250MGEA 41 EUR
BLDPBD314372-250MG BLDPBD314372-5G BLDPBD314372-100MG BLDPBD314372-1G
4-Butylthiobenzamide 95%
4-Butylthiobenzamide
Formula: C₁₁H₁₅NS
MW: 193,3085 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09025670
CAS Number: 1208077-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR