2-Bromo-2'-fluoro-4'-methoxyacetophenone 98%

Synonyms: 2-Bromo-1-(2-fluoro-4-methoxyphenyl)ethanone,2-Fluoro-4-methoxyphenacyl bromide

BD315348-1G BD315348-5G BD315348-250MG
BLDPBD315348-1GEA 75.6 EUR
BLDPBD315348-1G BLDPBD315348-5G BLDPBD315348-250MG
2-Bromo-2'-fluoro-4'-methoxyacetophenone 98%
2-Bromo-2'-fluoro-4'-methoxyacetophenone

Storage condition: Inert atmosphere, 2-8°C

Formula: C₉H₈BrFO₂
MW: 247,0600 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03730100
CAS Number: 157014-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR