Ethyl 1-methyl-1H-pyrrole-2-carboxylate 95%

BD315488-5G BD315488-250MG BD315488-100MG BD315488-1G
BLDPBD315488-5GEA 332 EUR
BLDPBD315488-5G BLDPBD315488-250MG BLDPBD315488-100MG BLDPBD315488-1G
Ethyl 1-methyl-1H-pyrrole-2-carboxylate 95%
Ethyl 1-methyl-1H-pyrrole-2-carboxylate
Formula: C₈H₁₁NO₂
MW: 153,178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02179573
CAS Number: 23466-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR