3-Bromo-6-chloroimidazo[1,2-b]pyridazine 97%

BLDPBD31807-25GEA 94.5 EUR
BLDPBD31807-25G BLDPBD31807-250MG BLDPBD31807-100G BLDPBD31807-10G BLDPBD31807-5G BLDPBD31807-1G BLDPBD31807-500G
3-Bromo-6-chloroimidazo[1,2-b]pyridazine 97%
3-Bromo-6-chloroimidazo[1,2-b]pyridazine

Storage condition: Inert atmosphere, 2-8°C

Formula: C₆H₃BrClN₃
MW: 232,47 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09027274
CAS Number: 13526-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR