2-Bromo-6-fluoro-4-nitroaniline 97%

Synonyms: 2-Bromo-6-Fluoro-4-nitroaniline

BLDPBD320525-10GEA 214 EUR
BLDPBD320525-10G BLDPBD320525-1G BLDPBD320525-250MG BLDPBD320525-5G
2-Bromo-6-fluoro-4-nitroaniline 97%
2-Bromo-6-fluoro-4-nitroaniline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₆H₄BrFN₂O₂
MW: 235,0100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09800780
CAS Number: 455-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR