4-(3-Hydroxypropylcarbamoyl)phenylboronic acid pinacol ester 96%

Supplier: BLD PHARMATECH GMBH

Synonyms: N-(3-Hydroxy-propyl)-4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-benzamideN-(3-Hydroxypropyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

BLDPBD321893-1GEA 81.9 EUR
BLDPBD321893-1G BLDPBD321893-250MG BLDPBD321893-5G
4-(3-Hydroxypropylcarbamoyl)phenylboronic acid pinacol ester 96%
4-(3-Hydroxypropylcarbamoyl)phenylboronic acid pinacol ester

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₆H₂₄BNO₄
MW: 305,1800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16294505
CAS Number: 1449134-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR