6-Bromoquinolin-2(1H)-one 96%
Supplier: BLD PHARMATECH GMBH
|
|
BLDPBD32338-10GEA
0
EUR
BLDPBD32338-10G
BLDPBD32338-5G
BLDPBD32338-250MG
BLDPBD32338-25G
BLDPBD32338-1G
BLDPBD32338-100G
6-Bromoquinolin-2(1H)-one 96%
6-Bromoquinolin-2(1H)-one
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formula:
C₉H₆BrNO MW: 224,05 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08703169 CAS Number: 1810-66-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...