2,2-Difluoro-2-(2-(trifluoromethoxy)phenyl)acetic acid 98%

BD324921-1G BD324921-100MG BD324921-250MG
BLDPBD324921-1GEA 248 EUR
BLDPBD324921-1G BLDPBD324921-100MG BLDPBD324921-250MG
2,2-Difluoro-2-(2-(trifluoromethoxy)phenyl)acetic acid 98%
2,2-Difluoro-2-(2-(trifluoromethoxy)phenyl)acetic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₅F₅O₃
MW: 256,1262 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11855928
CAS Number: 1133116-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR