4-(1-Piperidinyl)aniline 97%

Synonyms: 4-Piperidinoaniline

BD32786-5G BD32786-1G BD32786-250MG
BLDPBD32786-5GEA 36.8 EUR
BLDPBD32786-5G BLDPBD32786-1G BLDPBD32786-250MG
4-(1-Piperidinyl)aniline 97%
4-(1-Piperidinyl)aniline

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₁₁H₁₆N₂
MW: 176,26 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00051688
CAS Number: 2359-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR