Methyl 2-bromo-5-hydroxybenzoate 95%

Synonyms: Methyl 2-bromo-5-hydroxybenzenecarboxylate

BLDPBD328578-1GEA 114 EUR
BLDPBD328578-1G BLDPBD328578-100MG BLDPBD328578-5G BLDPBD328578-250MG
Methyl 2-bromo-5-hydroxybenzoate 95%
Methyl 2-bromo-5-hydroxybenzoate

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₇BrO₃
MW: 231,043 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10566797
CAS Number: 154607-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR