2,4-Dichlorobenzene-1,3-diol 95%

Supplier: BLD PHARMATECH GMBH

BLDPBD328596-5GEA 206 EUR
BLDPBD328596-5G BLDPBD328596-10G BLDPBD328596-1G BLDPBD328596-250MG
2,4-Dichlorobenzene-1,3-diol 95%
2,4-Dichlorobenzene-1,3-diol

Storage condition: Sealed in dry, 2-8°C

Formula: C₆H₄Cl₂O₂
MW: 179,001 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13152136
CAS Number: 16606-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR