Furan-2,5-dicarboxylic acid 98%
|
|
BLDPBD3299-5GEA
13.7
EUR
BLDPBD3299-5G
BLDPBD3299-500G
BLDPBD3299-100G
BLDPBD3299-1G
BLDPBD3299-25G
BLDPBD3299-0.025G
BLDPBD3299-10G
Furan-2,5-dicarboxylic acid 98%
Furan-2,5-dicarboxylic acid
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formula:
C₆H₄O₅ MW: 156,09 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00016582 CAS Number: 3238-40-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...