2-Iodo-p-toluic acid 95%

Synonyms: 2-Iodo-4-methylbenzoic acid

BD330595-25G BD330595-250MG BD330595-1G BD330595-100MG BD330595-5G
BLDPBD330595-25GEA 1230 EUR
BLDPBD330595-25G BLDPBD330595-250MG BLDPBD330595-1G BLDPBD330595-100MG BLDPBD330595-5G
2-Iodo-p-toluic acid 95%
2-Iodo-p-toluic acid
Formula: C₈H₇IO₂
MW: 262,0400 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01312232
CAS Number: 1829-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR