4-(Cyclopropylmethoxy)-benzoic acid 98%

Supplier: BLD PHARMATECH GMBH

Synonyms: 4-(Cyclopropylmethoxy)benzoic acid

BLDPBD331388-1GEA 303 EUR
BLDPBD331388-1G BLDPBD331388-100MG BLDPBD331388-250MG
4-(Cyclopropylmethoxy)-benzoic acid 98%
4-(Cyclopropylmethoxy)-benzoic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₁H₁₂O₃
MW: 192,211 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11156580
CAS Number: 355391-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR