1H-Pyrazole-4-carboxylic acid 98%

Synonyms: 4-Pyrazolecarboxylic acid

BD33461-500G BD33461-25G BD33461-1G BD33461-10G BD33461-5G BD33461-100G
BLDPBD33461-500GEA 906 EUR
BLDPBD33461-500G BLDPBD33461-25G BLDPBD33461-1G BLDPBD33461-10G BLDPBD33461-5G BLDPBD33461-100G
1H-Pyrazole-4-carboxylic acid 98%
1H-Pyrazole-4-carboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₄H₄N₂O₂
MW: 112,09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011558
CAS Number: 37718-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR