(R)-3-Aminobutyric acid 97%

Synonyms: (R)-3-Aminobutanoic acid

BD33462-10G BD33462-25G BD33462-5G BD33462-1G BD33462-100G BD33462-500G
BLDPBD33462-10GEA 13.7 EUR
BLDPBD33462-10G BLDPBD33462-25G BLDPBD33462-5G BLDPBD33462-1G BLDPBD33462-100G BLDPBD33462-500G
(R)-3-Aminobutyric acid 97%
(R)-3-Aminobutyric acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₄H₉NO₂
MW: 103,1198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00211284
CAS Number: 3775-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR