6-Iodoisoquinoline 95%

Supplier: BLD PHARMATECH GMBH

BLDPBD335296-100MGEA 107 EUR
BLDPBD335296-100MG BLDPBD335296-5G BLDPBD335296-1G BLDPBD335296-250MG
6-Iodoisoquinoline 95%
6-Iodoisoquinoline

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₉H₆IN
MW: 255,055 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11109663
CAS Number: 75476-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR