3-(Methylsulphonyl)benzaldehyde 95%

Synonyms: 3-Formylphenyl methyl sulphone

BLDPBD33675-250MGEA 41 EUR
BLDPBD33675-250MG BLDPBD33675-1G BLDPBD33675-5G BLDPBD33675-100MG
3-(Methylsulphonyl)benzaldehyde 95%
3-(Methylsulphonyl)benzaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formula: C₈H₈O₃S
MW: 184,212 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11052329
CAS Number: 43114-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR