2-Chloro-9-methyl-7H-purin-8(9H)-one 95%

Supplier: BLD PHARMATECH GMBH

BLDPBD337896-250MGEA 275 EUR
BLDPBD337896-250MG BLDPBD337896-100MG BLDPBD337896-1G
2-Chloro-9-methyl-7H-purin-8(9H)-one 95%
2-Chloro-9-methyl-7H-purin-8(9H)-one

Storage condition: Inert atmosphere, 2-8°C

Formula: C₆H₅ClN₄O
MW: 184,583 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24567184
CAS Number: 1273315-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR