1-Cyclopropyl-1-(4-methoxyphenyl)methylamine 97%

Supplier: BLD PHARMATECH GMBH

BLDPBD338300-100MGEA 149 EUR
BLDPBD338300-100MG BLDPBD338300-250MG BLDPBD338300-1G
1-Cyclopropyl-1-(4-methoxyphenyl)methylamine 97%
1-Cyclopropyl-1-(4-methoxyphenyl)methylamine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₁H₁₅NO
MW: 177,243 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00044830
CAS Number: 54398-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR