2-Amino-1-isopropylbenzimidazole 95%

Supplier: BLD PHARMATECH GMBH

BLDPBD338509-100MGEA 30.5 EUR
BLDPBD338509-100MG BLDPBD338509-1G BLDPBD338509-250MG
2-Amino-1-isopropylbenzimidazole 95%
2-Amino-1-isopropylbenzimidazole

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₀H₁₃N₃
MW: 175,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00159972
CAS Number: 90871-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR