Diethyl 3-(tert-Butyldimethylsilyloxy)glutarate 95%

Supplier: BLD PHARMATECH GMBH

BLDPBD338512-5GEA 107 EUR
BLDPBD338512-5G BLDPBD338512-25G BLDPBD338512-1G BLDPBD338512-10G BLDPBD338512-250MG
Diethyl 3-(tert-Butyldimethylsilyloxy)glutarate 95%
Diethyl 3-(tert-Butyldimethylsilyloxy)glutarate

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₅H₃₀O₅Si
MW: 318,481 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16251279
CAS Number: 91424-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR