5-Amino-3-bromo-2-methoxypyridine 98%
Supplier: BLD PHARMATECH GMBH
|
|
Synonyms:
5-Bromo-6-methoxypyridin-3-amine
BLDPBD33974-1GEA
9.5
EUR
BLDPBD33974-1G
BLDPBD33974-5G
BLDPBD33974-100G
BLDPBD33974-25G
BLDPBD33974-250MG
BLDPBD33974-10G
5-Amino-3-bromo-2-methoxypyridine 98%
5-Amino-3-bromo-2-methoxypyridine
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
C₆H₇BrN₂O MW: 203,0400 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06659518 CAS Number: 53242-18-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...