4-Bromo-1-naphthol 97%

Synonyms: 4-Bromonaphthalen-1-ol

BD34131-5G BD34131-25G BD34131-250MG BD34131-1G BD34131-10G BD34131-100G
BLDPBD34131-5GEA 22.1 EUR
BLDPBD34131-5G BLDPBD34131-25G BLDPBD34131-250MG BLDPBD34131-1G BLDPBD34131-10G BLDPBD34131-100G
4-Bromo-1-naphthol 97%
4-Bromo-1-naphthol

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₇BrO
MW: 223,0700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03129265
CAS Number: 571-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR