Methyl-2-pyrazinecarboxylate 97%

Supplier: BLD PHARMATECH GMBH

Synonyms: Methyl pyrazine-2-carboxylateMethyl 2-pyrazinecarboxylate

BLDPBD3516-10GEA 25.2 EUR
BLDPBD3516-10G BLDPBD3516-1G BLDPBD3516-5G BLDPBD3516-25G BLDPBD3516-500G BLDPBD3516-100G
Methyl-2-pyrazinecarboxylate 97%
Methyl 2-pyrazinecarboxylate

Storage condition: Sealed in dry, Room Temperature

Formula: C₆H₆N₂O₂
MW: 138,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00014611
CAS Number: 6164-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR