4-Bromo-1,3-dimethyl-1H-pyrazole-5-carboxylic acid 96%

BLDPBD3590-1GEA 136 EUR
BLDPBD3590-1G BLDPBD3590-5G BLDPBD3590-250MG
4-Bromo-1,3-dimethyl-1H-pyrazole-5-carboxylic acid 96%
4-Bromo-1,3-dimethyl-1H-pyrazole-5-carboxylic acid

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formula: C₆H₇BrN₂O₂
MW: 219,0360 g/mol
Storage Temperature: Freezer
MDL Number: MFCD02090873
CAS Number: 5775-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR